missing translation for 'onlineSavingsMsg'
Learn More

Dynactin p50 Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611003

 View more versions of this product

Product Code. 15825759

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Dyneins are ubiquitous, multimeric proteins that are responsible for minus end-directed microtubule-based organelle transport. They are divided into two classes, cytosolic and axonemal. The activity of cytosolic dynein is regulated by the multiprotein dynactin complex with which it co-purifies. Dynactin is composed of at least nine polypeptides with p50 being the second most abundant subunit. The complex consists of an F-actin-like core filament of actin-related protein Arp1, a heterodimer that acts as a cap at one end, and a 62kDa subunit at the other end. p150[glued] is the largest subunit of the complex. The precise function of dynactin remains to be determined, but it may be recruited to prometaphase kinetochores to assist in alignment and spindle organization during mitosis. Additionally, dynactin localizes at the tail of cytosolic dynein and activates dynein-mediated vesicle movement. Therefore, dynactin may function to anchor or target dynein to the kinetochore during mitosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dynactin p50
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Dynactin p50 aa. 55-196
150 μg
Cell Biology
Drosophila, Human, Mouse, Rat
IgG1
Western Blot
25
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.