Learn More
DYKDDDDK Tag Monoclonal Antibody (FG4R), HRP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191878HRP
217.55 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
similar epitope as the FLAG tag from Sigma MA1-91878 detects Flag tag from various samples. MA1-91878 has been successfully used in Western blot applications. The MA1-91878 immunogen is synthetic peptide (DYKDDDDK) coupled to KLH.
DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for Western Blot.Specifications
DYKDDDDK Tag | |
Monoclonal | |
1 mg/mL | |
antibody with 0.07% Kathon | |
Mouse | |
Protein A | |
RUO | |
4° C | |
Liquid | |
Tag |
Western Blot | |
FG4R | |
HRP | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
Synthetic peptide (DYKDDDDK) coupled to KLH. | |
50 μL | |
Primary | |
Antibody | |
IgG2b |
Safety and Handling
- DYKDDDDK Tag Monoclonal Antibody (FG4R), HRP
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one