Learn More
DTNB, Mouse, Clone: 1D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DTNB.
Brand: Abnova H00001838-M03.100ug
Description
This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: MIEESGNKRKTMAEKRQLFIEMRAQNFDVIRLSTYRTACKLRFVQKRCNLHLVDIWNMIEAFRDNGLNTLDHTTEISVSRLETVISSIYSpecifications
| DTNB | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_183361 | |
| DTNB | |
| DTNB (NP_899205, 1 a.a. ∼ 89 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 1D3 | |
| Mouse monoclonal antibody raised against a partial recombinant DTNB. | |
| DTNB | |
| MGC17163/MGC57126 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1838 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.