Learn More
DsRed with w/control Mouse, Unlabeled, Clone: E64-1077, BD
Mouse Monoclonal Antibody with Control
Brand: BD Biosciences 551814
Description
DsRed Control Lysate, DsRed is naturally occurring fluorescent protein from a coral of the genus Discosoma. DsRed, like green fluorescent protein (GFP) can be introduced into either cultured cells or in transgenic animals to study the localization of a specific protein. It can also be used in fusion constructs with a protein of interest to examine protein-protein interactions or as a general marker. The crystal structure of DsRed has been solved and shown that DsRed monomers exhibit similar topology to GFP, but have additional chemical modifications which account for differences in the spectra, when compared to GFP. DsRed has a longer emission/absorption wavelength than GFP. DsRed migrates at 31kDa in SDS/PAGE. The antibody recognizes three forms of DsRed; 1, 2 and pTimer. A recombinant protein representing DsRed1 was used as the immunogen.
Host Species: Mouse
Clone: E64-1077
Isotype: IgG1 κ
Immunogen: DsRed1 recombinant protein
Formula Weight [Chemical]: 31kDa
Western Blotting
Specifications
| DsRed with w/control | |
| Monoclonal | |
| 0.25mg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| DsRed1 recombinant protein | |
| 50 μg | |
| Cell Biology | |
| Store undiluted at -20°C. |
| Western Blot | |
| E64-1077 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.