Learn More
Drp1 (Density Regulated Protein-1) Mouse, Unlabeled, Clone: 22, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611739
Description
In culture, cell density can have profound effects on gene expression, enzymatic activity, and cell signaling pathways. Using differential screening of cDNAs from low-passage nontumorigenic teratocarcinoma cells versus high passage tumorigenic cells, a protein was identified that is regulated in cell-density dependent manner. This protein, density-regulated protein 1 (drp1), contains putative sites for N-myristoylation and phosphorylation sites for cAMP and/or cGMP-dependent kinase, casein kinase II, and PKC. The expression of drp1 is enriched in high density cultures of both nontumorigenic and tumorigenic cell lines and is widely detected in adult organs, especially skeletal and cardiac muscle. In addition, increased expression of drp1 is not due to growth arrest as a result of serum starvation or TGF-β treatment nor is it a result of factors found in the media of high density cultures. Interestingly, drp1 is expressed highest in skeletal and cardiac muscle where unique cell-cell contacts are involved in muscle cell membrane depolarization and contraction. Thus, drp1 expression may be regulated by signaling pathways related to specific types of cell-cell contacts.
Host Species: Mouse
Clone: 22
Isotype: IgG2a
Species Reactivity [for Features Main]: Human
Immunogen: Human Drp1 aa. 19-201
Immunofluorescence, Western Blotting
Specifications
| Drp1 (Density Regulated Protein-1) | |
| Monoclonal | |
| 250μg/mL | |
| Density Regulated Protein-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 22 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human Drp1 aa. 19-201 | |
| 150 μg | |
| Cell Biology | |
| Canine, Human, Mouse, Rat | |
| IgG2a |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.