Learn More
Dopamine beta Hydroxylase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534664
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: SH-SY5Y. Predicted reactivity: Rhesus Monkey (86%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide.Specifications
Dopamine beta Hydroxylase | |
Polyclonal | |
Unconjugated | |
DBH | |
Dbh; DBM; dopamine beta hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor; dopamine beta-hydroxylase precursor (EC 1.14.17.1); dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); Dopamine-Beta-hydroxylase; DOPBHY; mixed function oxidase; Soluble dopamine beta-hydroxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13166, 1621, 25699 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P09172, Q05754, Q64237 | |
DBH | |
Recombinant fragment corresponding to a region within amino acids 8 and 320 of Dopamine beta Hydroxylase (Uniprot ID#P09172) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |