Learn More
tRNA aspartic acid methyltransferase 1, Mouse, Clone: 1E12, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant DNMT2.
Brand: Abnova H00001787-M01.100ug
Description
This gene encodes a protein responsible for the methylation of aspartic acid transfer RNA. While similar in sequence and structure to DNA cytosine methyltransferases, this gene is distinct and highly conserved in its function among taxa. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFHHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVEEKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYESpecifications
| tRNA aspartic acid methyltransferase 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC047733 | |
| TRDMT1 | |
| DNMT2 (AAH47733, 1 a.a. ∼ 391 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1E12 | |
| Mouse monoclonal antibody raised against a full length recombinant DNMT2. | |
| TRDMT1 | |
| DNMT2/M.HsaIIP/PuMet/RNMT1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 1787 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 λ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.