Learn More
dynein, axonemal, light intermediate chain 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human DNALI1 protein.
Brand: Abnova H00007802-B01.50uL
Description
This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects. [provided by RefSeq
Sequence: MIPPADSLLKYDTPVLVSRNTEKRSPKARLLKVSPQQPGPSGSAPQPPKTKLPSTPCVPDPTKQAEEILNAILPPREWVEDTQLWIQQVSSTPSTRMDVVHLQEQLDLKLQQRQARETGICPVRRELYSQCFDELIREVTINCAERGLLLLRVRDEIRMTIAAYQTLYESSVAFGMRKALQAEQGKSDMERKIAELETEKRDLERQVNEQKAKCEATEKRESERRQVEEKKHNEEIQFLKRTNQQLKAQLEGIIAPKKSpecifications
| dynein, axonemal, light intermediate chain 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human DNALI1 protein. | |
| DNALI1 | |
| P28/dJ423B22.5/hp28 | |
| Mouse | |
| 50 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Serum |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| BC046117.1 | |
| DNALI1 | |
| DNALI1 (AAH46117.1, 1 a.a. ∼ 258 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 7802 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.