Learn More
dynein, axonemal, light intermediate chain 1, Mouse, Clone: 2H3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant DNALI1.
Brand: Abnova H00007802-M04A.200uL
Description
This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects. [provided by RefSeq
Sequence: MIPPADSLLKYDTPVLVSRNTEKRSPKARLLKVSPQQPGPSGSAPQPPKTKLPSTPCVPDPTKQAEEILNAILPPREWVEDTQLWIQQVSSTPSTRMDVVHLQEQLDLKLQQRQARETGICPVRRELYSQCFDELIREVTINCAERGLLLLRVRDEIRMTIAAYQTLYESSVAFGMRKALQAEQGKSDMERKIAELETEKRDLERQVNEQKAKCEATEKRESERRQVEEKKHNEEIQFLKRTNQQLKAQLEGIIAPKKSpecifications
| dynein, axonemal, light intermediate chain 1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC046117 | |
| DNALI1 | |
| DNALI1 (AAH46117, 1 a.a. ∼ 258 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Yes | |
| 7802 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Immunoprecipitation, Western Blot | |
| 2H3 | |
| Mouse monoclonal antibody raised against a full-length recombinant DNALI1. | |
| DNALI1 | |
| P28/dJ423B22.5/hp28 | |
| Mouse | |
| 200 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.