Learn More
anti-DNA PKcs (phospho S2056), Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB18192-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Human. Predicted to work with Mouse, Rat
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 2050 - 2150 of Human DNA PKcs.
Applications:
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
| DNA PKcs (phospho S2056) | |
| 0.4-0.7mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Epigenetics And Nuclear Signaling | |
| Human, Mouse, Rat |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 2050 - 2150 of Human DNA PKcs. | |
| 100 μg | |
| Primary | |
| IgG |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.