missing translation for 'onlineSavingsMsg'
Learn More

DNA-PKcs Monoclonal Antibody, Clone: 18-2 + 25-4 + 42-psc, Invitrogen™

Product Code. 11324843
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324843 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11324843 Supplier Invitrogen Supplier No. MA513404

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

DNA-PKcs Antibody (18-2 + 25-4 + 42-psc), MA5-13404, from Invitrogen™. Species Reactivity: Ck, Hm, Hu, Ms, Xnl; Applications:GS, IF, IHC (P), IP, WB.

DNA-PKcs Antibody (18-2 + 25-4 + 42-psc), MA5-13404, from Invitrogen. Species Reactivity: Chicken, Hamster, Human, Mouse, Xenopus laevis; Applications: Gel Shift, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks
Host Species: Mouse
Cross Reactivity: Chicken, Hamster, Human, Mouse, Xenopus laevis
Isotype: IgG1/IgG2a
Clone: 18-2 + 25-4 + 42-psc
Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells
Formulation: 500μL purified antibody (0.2mg/mL) in PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Synonyms: DNA-PK catalytic subunit, DNA-dependent protein kinase catalytic subunit, hyper-radiosensitivity of murine scid mutation, complementing 1, p460
Uniprot Accession: P78527, P97313, Q8QGX4, Q9DEI1
Entrez GeneID: 100770748, 19090, 373602, 395376, 5591
Gene Symbol: PRKDC
MA5-13404 targets DNA-PKcs in gel shift, immunofluorescence, immunohistochemistry (paraffin), immunoprecipitation, and western blot applications and shows reactivity with Chicken, Hamster, Human, mouse, and Xenopus laevis samples. The MA5-13404 immunogen is human DNA-dependent Protein Kinase purified from HeLa cells.
References:
Test: ICC, Species: Hu, Reference: Subnuclear localization of Ku protein: functional association with RNA polymerase II elongation sites. (2002 Nov) Mol Cell Biol, Vol: 22, 22, 8088-99, Mo X, Dynan WS; Pubmed ID: 12391174
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA-PKcs
Applications Gel Shift, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 18-2 + 25-4 + 42-psc
Conjugate Unconjugated
Formulation 500μL purified antibody (0.2mg/mL) in PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Accession No. P78527, P97313, Q8QGX4, Q9DEI1
Gene Alias DNA-PK catalytic subunit, DNA-dependent protein kinase catalytic subunit, hyper-radiosensitivity of murine scid mutation, complementing 1, p460
Gene Symbols PRKDC
Host Species Mouse
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100770748, 19090, 373602, 395376, 5591
Target Species Chicken, Hamster, Human, Mouse, X. laevis
Content And Storage Store at 4°C
Form Purified
Isotype IgG1, IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.