missing translation for 'onlineSavingsMsg'
Learn More

DMRT2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA541694

Product Code. 15554962

  • £419.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: DPQAGSAAGD WEIDVESLEL EEDVCGAPRS TPPGPSPPPA DGDCEDDEDD Sequence homology: Human: 100%; Rabbit: 86%

DMRT2 Polyclonal antibody specifically detects DMRT2 in Human samples. It is validated for Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DMRT2
Polyclonal
Unconjugated
DMRT2
dmrt2; doublesex and mab-3 related transcription factor 2; doublesex- and mab-3-related transcription factor 2; Doublesex-like 2 protein; doublesex-like 2 protein (DSXL-2); DSXL2; DSXL-2; LOW QUALITY PROTEIN: doublesex- and mab-3-related transcription factor 2; Terra; terra homolog; terra like protein antibody; terra-like protein
Rabbit
Protein A
RUO
10655
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Y5R5
DMRT2
Synthetic peptide directed towards the N-terminal of human DMRT2
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.