missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO3 Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522886
This item is not returnable.
View return policy
Description
The target sequence has 93% sequence homology with hamster. Suggested positive control: rat placenta tissue lysate.
Dio3, or type 3 iodothyronine deiodinase, can regulate circulating fetal thyroid hormone concentrations throughout gestation. It has an essential role for regulation of thyroid hormone inactivation during embryological development, and may play a role in preventing premature exposure of developing fetal tissues to adult levels of thyroid hormones. Dio3 has been linked to the development of consumptive hypothyroidism in both infants and adults.
Specifications
| DIO3 | |
| Polyclonal | |
| Unconjugated | |
| P55073, Q91ZI8, P49897 | |
| DIO3 | |
| Synthetic peptide made to an internal portion of the rat protein (within residues 1-100). | |
| 100 μL | |
| Primary | |
| Hamster, Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| DIO3 | |
| 5DIII, D3, DIOIII, TXDI3, iodothyronine deiodinase, placental type, thyroxine deiodinase type III (selenoprotein), type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5′ deiodinase, type-III 5′-deiodinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100753739, 107585, 1735, 29475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction