missing translation for 'onlineSavingsMsg'
Learn More

DGAT2 Polyclonal Antibody, Invitrogen™

Product Code. 13269918
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13269918 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13269918 Supplier Invitrogen Supplier No. PA521722

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

DGAT2 Antibody, PA5-21722, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry (Paraffin), Western Blot.

DGAT2 Antibody, PA5-21722, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry (Paraffin), Western Blot.


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Recombinant fragment corresponding to a region within amino acids 153 and 388 of DGAT2 (Uniprot ID#Q96PD7)
Formulation: 100μL purified antibody (1mg/mL) in PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Synonyms: HMFN1045, GS1999FULL, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase-like protein 2, diglyceride acyltransferase 2
Uniprot Accession: Q5FVP8, Q96PD7, Q9DCV3
Entrez GeneID: 252900, 67800, 84649
Gene Symbol: DGAT2
PA5-21722 targets DGAT2 in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-21722 immunogen is a recombinant fragment corresponding to a region within amino acids 153 and 388 of DGAT2 (Uniprot ID#Q96PD7).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGAT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Accession No. Q5FVP8, Q96PD7, Q9DCV3
Gene Alias HMFN1045, GS1999FULL, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase-like protein 2, diglyceride acyltransferase 2
Gene Symbols DGAT2
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 153 and 388 of DGAT2 (Uniprot ID#Q96PD7)
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252900, 67800, 84649
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.