missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DENND1C Polyclonal Antibody
GREENER_CHOICE

Product Code. 15567072
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15567072 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15567072 Supplier Invitrogen™ Supplier No. PA546140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: PSSLEDPRAR PPKALLAERA HLQPREEPGA LNSPATPTSN CQKSQPSSRP Sequence homology: Human: 100%.

Clathrin-mediated endocytosis is a major mechanism for internalization of proteins and lipids. Members of the connecdenn family, such as DENND1C, function as guanine nucleotide exchange factors (GEFs) for the early endosomal small GTPase RAB35 and bind to clathrin and clathrin adaptor protein-2. Thus, connecdenns link RAB35 activation with the clathrin machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND1C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DENND1C
Gene Accession No. Q8IV53
Gene Alias Connecdenn 3; DENN domain containing 1C; DENN domain-containing protein 1C; DENN/MADD domain containing 1C; DENND1C; FAM31C; family with sequence similarity 31, member C; Protein FAM31C
Gene Symbols DENND1C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DENND1C (aa 741-790).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79958
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.