Learn More
DEFB4, Mouse, Clone: 4C4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DEFB4.
Brand: Abnova H00001673-M01.100ug
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 4, an antibiotic peptide which is locally regulated by inflammation. [provided by RefSeq
Sequence: GIGDPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKPSpecifications
| DEFB4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004942 | |
| DEFB4 | |
| DEFB4 (NP_004933, 24 a.a. ∼ 64 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 4C4 | |
| Mouse monoclonal antibody raised against a partial recombinant DEFB4. | |
| DEFB4 | |
| DEFB-2/DEFB102/DEFB2/HBD-2/SAP1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1673 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Safety and Handling
- DEFB4 monoclonal antibody (M01)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodum chloride, sodium phopshate dibasic, potassium phosphate monobasic, potassium chloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.