missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX59 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531732
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, Neuro2A, GL261, C8D30. Predicted reactivity: Mouse (90%), Rat (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DDX59 is a protein coding gene.
Specifications
| DDX59 | |
| Polyclonal | |
| Unconjugated | |
| DDX59 | |
| 1210002B07Rik; 4833411G06Rik; DDX59; DEAD (Asp-Glu-Ala-Asp) box polypeptide 59; DEAD box protein 59; DEAD-box helicase 59; OFD5; Probable ATP-dependent RNA helicase DDX59; zinc finger HIT domain-containing protein 5; Znhit5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 67997, 83479 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q5T1V6, Q9DBN9 | |
| DDX59 | |
| Recombinant protein encompassing a sequence within the center region of human DDX59. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction