Learn More
DEAD (Asp-Glu-Ala-Asp) box polypeptide 50, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human DDX50 protein.
Brand: Abnova H00079009-B01.50uL
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box enzyme that may be involved in ribosomal RNA synthesis or processing. This gene and DDX21, also called RH-II/GuA, have similar genomic structures and are in tandem orientation on chromosome 10, suggesting that the two genes arose by gene duplication in evolution. This gene has pseudogenes on chromosomes 2, 3 and 4. Alternative splicing of this gene generates multiple transcript variants, but the full length nature of all the other variants but one has not been defined. [provided by RefSeq
Sequence: MPGKLLWGDIMELEAPLEESESQKKERQKSDRRKSRHHYDSDEKSETRENGVTDDLDAPKAKKSKMKEKLNGDTEEGFNRLSDEFSKSHKSRRKDLPNGDIDEYEKKSKRVSSLDTSTHKSSDNKLEETLTREQKEGAFSNFPISEETIKLLKGRGVTYLFPIQVKTFGPVYEGKDLIAQARTGTGKTFSFAIPLIERLQRNQETIKKSRSPKVLVLAPTRELANQVAKDFKDITRKLSVACFYGGTSYQSQINHIRNGIDILVGTPGRIKDHLQSGRLDLSKLRHVVLDEVDQMLDLGFAEQVEDIIHESYKTDSEDNPQTLLFSATCPQWVYKVAKKYMKSRYEQVDLVGKMTQKAATTVEHLAIQCHWSQRPAVIGDVLQVYSGSEGRAIIFCETKKNVTEMAMNPHIKQNAQCLHGDIAQSQREITLKGFREGSFKVLVATNVAARGLDIPEVDLVIQSSPPQDVESYIHRSGRTGRAGRTGICICFYQPRERGQLRYVEQKAGITFKRVGVPSTMDLVKSKSMDAIRSLASVSYAAVDFFRPSAQRLIEEKGAVDALAAALAHISGASSFEPRSLITSDKGFVTMTLESLEEIQDVSCAWKELNRKLSSNAVSQITRMCLLKGNMGVCFDVPTTESERLQAEWHDSDWILSVPAKLPEIEEYYDGNTSSNSRQRSGWSSGRSGRSGRSGGRSGGRSGRQSRQGSRSGSRQDGRRRSGNRNRSRSGGHKRSFDSpecifications
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 50 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human DDX50 protein. | |
| DDX50 | |
| GU2/GUB/MGC3199/RH-II/GuB | |
| Mouse | |
| 50 μL | |
| Yes | |
| 79009 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_024045.1 | |
| DDX50 | |
| DDX50 (NP_076950.1, 1 a.a. ∼ 737 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.