Learn More
DCP1 decapping enzyme homolog A (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DCP1A.
Brand: Abnova H00055802-A01.50uL
Description
Decapping is a key step in general and regulated mRNA decay. The protein encoded by this gene is a decapping enzyme. This protein and another decapping enzyme form a decapping complex, which interacts with the nonsense-mediated decay factor hUpf1 and may be recruited to mRNAs containing premature termination codons. This protein also participates in the TGF-beta signaling pathway. [provided by RefSeq
Sequence: STQLSNLGSTETLEEMPSGSQDKSAPSGHKHLTVEELFGTSLPKEQPAVVGLDSEEMERLPGDASQKEPNSFLPFPFEQLGGAPQSETLGVPSAAHHSVQSpecifications
| DCP1 decapping enzyme homolog A (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant DCP1A. | |
| DCP1A | |
| FLJ21691/HSA275986/Nbla00360/SMAD4IP1/SMIF | |
| Mouse | |
| 50 μL | |
| Yes | |
| 55802 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_018403 | |
| DCP1A | |
| DCP1A (NP_060873, 186 a.a. ∼ 285 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.