Learn More
DCK, Mouse, Clone: 2F10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DCK.
Brand: Abnova H00001633-M03.100ug
Description
Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity. [provided by RefSeq
Sequence: WMNNQFGQSLELDGIIYLQATPETCLHRIYLRGRNEEQGIPLEYLEKLHYKHESWLLHRTLKTNFDYLQEVPILTLDVNEDFKDKYESLVEKVKEFLSTLSpecifications
| DCK | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000788 | |
| DCK | |
| DCK (NP_000779, 161 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunofluorescence, Western Blot | |
| 2F10 | |
| Mouse monoclonal antibody raised against a partial recombinant DCK. | |
| DCK | |
| MGC117410/MGC138632 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1633 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.