Learn More
DCIR Mouse anti-Human, Alexa Fluor 647, Clone: I3-612, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558220
Description
The I3-612 antibody reacts with DCIR, or dendritic cell immunoreceptor, a newly identified member of the calcium-dependent (C-type) lectin family. DCIR is a type II membrane glycoprotein with a single carbohydrate recognition domain (CRD). Its cytoplasmic tail contains a consensus immunoreceptor tyrosine-based inhibitory motif (ITIM). It is expressed on monocytes, macrophages, B-lymphocytes, granulocytes, but not on NK or T cells. DCIR is strongly expressed on dendritic cells derived from peripheral blood monocytes cultured in presence of GM-CSF and IL-4. Down-regulation is observed on these with further maturation using TNF-α or LPS. Reports describe the detection of two distinct inhibitory signals depending on its ITIM tyrosine residue.
Host Species: Mouse
Clone: I3-612
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
| DCIR | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| I3-612 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.