missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ubiquitin domain containing 2, Mouse, Clone: 1D3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant DC-UbP.
Brand: Abnova H00092181-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MTDGQLRSKRDEFWDTAPAFEGRKEIWDALKAAAHAFESNDHELAQAIIDGANITLPHGALTECYDELGNRYQLPVYCLAPPINMIEEKSDIETLDIPEPPPNSGYECQLRLRLSTGKDLKLVVRSTDTVFHMKRRLHAAEGVEPGSQRWFFSGRPLTDKMKFEELKIPKDYVVQVIVSQPVQNPTPVENSpecifications
| ubiquitin domain containing 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC019910 | |
| UBTD2 | |
| DC-UbP (AAH19910, 1 a.a. ∼ 190 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 92181 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1D3 | |
| Mouse monoclonal antibody raised against a full length recombinant DC-UbP. | |
| UBTD2 | |
| DC-UbP/DCUBP/MGC30022 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction