Learn More
Cytokeratin Mouse anti-Human, FITC, Clone: CAM 5.2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 347653
Description
Anti-Cytokeratin (CAM 5.2) reagent has a primary reactivity with human keratin proteins that correspond to Moll's peptides No.7 and No.8, Mr 48 and 52kDa, respectively.Cytokeratin 7 and 8 are present on secretory epithelia of normal human tissue but not on stratified squamous epithelium. Anti-Cytokeratin (CAM 5.2) stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. Anti-Cytokeratin (CAM 5.2) may not react with some squamous cell carcinomas.
Flow Cytometry, Intracellular Staining
Specifications
| Cytokeratin | |
| Monoclonal | |
| 25μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human HT29 colorectal carcinoma cell line | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| CAM 5.2 | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.