missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hair Cortex Cytokeratin Monoclonal Antibody (AE13)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA15765

Product Code. 11554353

  • £520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-5765 detects Cytokeratin, AE13 in human and mouse samples. MA1-5765 has been successfully used in Western blot, immunocytochemistry, immunofluorescence and immunohistochemistry procedures. The MA1-5765 immunogen is human hair keratins. This antibody reacts with acidic 44-46 kDa hair cortex keratins.

Cytokeratins, a group comprising at least 29 different proteins, are characteristic of epithelial and trichocytic cells. Cytokeratin 5, 6 and 8 are members of the type II neutral-to-basic subfamily. Monoclonal antibodies to cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification. PCK-26 facilitates the typing of normal, metaplastic and neoplastic cells and it may aid in the discrimination of carcinomas and non-epithelial tumors such as sarcomas, lymphomas and neural tumors. It is also useful in detecting micro-metastases in lymph nodes and other tissues and for determining the origin of poorly differentiated tumors (1,2).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hair Cortex Cytokeratin
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Q6A162, Q6IFX3
KRT40
Human hair keratins from P3X63Ag. 8.653 cells.
100 μL
Primary
Human, Mouse
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Western Blot
AE13
Unconjugated
KRT40
CK-40; Cytokeratin-40; K40; KA36; keratin 40; keratin 40, type I; keratin, type I cytoskeletal 40; Keratin-40; Krt40; ortholog of type I hair keratin KA36; type I hair keratin KA36
Mouse
Protein G
RUO
125115, 406221
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.