Learn More
Invitrogen™ Cytokeratin 15 Monoclonal Antibody (LHK15)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA511344
Product Code. 11304433
  • £402.00 / 500µL
Description
MA5-11344 targets Cytokeratin 15 in ICC/IF, IHC-P and Western blot applications and shows reactivity with Bovine, Human, mouse, Non-human primate and Rat samples. The MA5-11344 immunogen is a 17-mer synthetic peptide from the C-terminus of human cytokeratin 15.
Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21. 2.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 15
Monoclonal
0.2 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4
P19012, Q61414, Q6IFV3
KRT15
A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15.
500 μL
Primary
Bovine, Human, Mouse, Rat
Antibody
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
LHK15
Unconjugated
KRT15
AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012}
Mouse
Protein A/G
RUO
16665, 287700, 3866, 507464
4°C