missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP11B1 Polyclonal Antibody

Description
Peptide sequence: ARNPNVQQAL RQESLAAAAS ISEHPQKATT ELPLLRAALK ETLRLYPVGL Sequence homology: Human: 100%; Pig: 79%; Sheep: 79%.
The steroid 11beta-hydroxylase gene, also designated Cyp11b-1, is a marker for the functional differentiation of cells in the zonae fasciculata reticularis. The deduced protein CYP11B1 consists of 466 amino acids containing a secretory signal, epidermal growth factor-like repeats, and a proteolytically inactive cathepsin B-related sequence. A related protein, human aldosterone synthase (CYP11B2), is involved in substrate recognition and conversion, with a functionally significant residue 112 in the N-terminal region of human CYP11B2. The inherited disorder glucocorticoid-remediable aldosteronism is caused by a chimeric gene duplication between the CYP11B1 and CYP11B2 genes. This disorder is characterized by hyperaldosteronism and high levels of 18-hydroxycortisol and 18-oxocortisol, which are under ACTH control.
Specifications
Specifications
| Antigen | CYP11B1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | CYP11B1 |
| Gene Accession No. | P15538 |
| Gene Alias | 11-beta-hydroxylase; AA389061; aldosterone synthase; Cp11ba; CPN1; CYP11B; CYP11B1; Cyp11b-1; CYPXIB1; Cytochrome P450 11B1, mitochondrial; cytochrome P450 family 11 subfamily B member 1; cytochrome p450 XIB1; cytochrome P450(11 beta)-DS; cytochrome P450, family 11, subfamily b, polypeptide 1; cytochrome P450, subfamily 11B, polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; Cytochrome P450, subfamily XIB, polypeptide 1 (steroid 11-beta-hydroxylase); cytochrome P450C11; cytochrome P-450c11; DKFZp686B05283; FHI; FLJ36771; P450(11 beta)-DS; P450C 11; P450C11; RATCP11BA; S11BH; steroid 11-beta-hydroxylase; steroid 11-beta-hydroxylase, CYP11B1; steroid 11-beta-monooxygenase; steroid-11-beta-hydroxylase |
| Gene Symbols | CYP11B1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?