Learn More
Cytochrome c Mouse anti-Human, Alexa Fluor 488, Clone: 6H2.B4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560263
Description
A cytochrome is an electron-transporting protein that contains a heme prosthetic group. Cytochromes have been known to be essential components of the mitochondrial respiratory chain since 1925. The iron atom of the heme group in cytochromes alternates between a reduced ferrous (+2) state and an oxidized ferric (+3) state during electron transport in oxidative phosphorylation. Cytochromes are classified into four groups (a, b, c and d) according to spectrochemical characteristics, and there are five cytochromes between coenzyme QH2 and O2 in the electron transport chain. Cytochrome c is a water-soluble protein that either promotes cell survival or death, depending upon its intracellular location. In healthy cells, it is a peripheral membrane protein of the mitochondria that transports electrons from the coenzyme QH2 cytochrome c reductase complex to the cytochrome c oxidase complex. When proapoptotic stimuli induce breakdown of the mitochondria, cytochrome c is released to the cytosol where it functions in the activation of caspases that trigger apoptosis. The 6H2.B4 monoclonal antibody has been reported to recognize the native and not the denatured form of rat, mouse, and human cytochrome c. Furthermore, studies utilizing competitive ELISA indicate that mAb 6H2.B4 binds to a region around residue 62 of rat cytochrome c.
Host Species: Mouse
Clone: 6H2.B4
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Rat cytochrome c
Bioimaging
Specifications
| Cytochrome c | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| In vitro Assay | |
| 6H2.B4 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Rat cytochrome c | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.