Learn More
cylindromatosis (turban tumor syndrome), Mouse, Clone: 3A9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CYLD.
Brand: Abnova H00001540-M02.100ug
Description
This gene is encodes a cytoplasmic protein with three cytoskeletal-associated protein-glycine-conserved (CAP-GLY) domains that functions as a deubiquitinating enzyme. Mutations in this gene have been associated with cylindromatosis, multiple familial trichoepithelioma, and Brooke-Spiegler syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: QNMELFAVLCIETSHYVAFVKYGKDDSAWLFFDSMADRDGGQNGFNIPQVTPCPEVGEYLKMSLEDLHSLDSRRIQGCARRLLCDAYMCMYQSPTMSLYKSpecifications
| cylindromatosis (turban tumor syndrome) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC012342 | |
| CYLD | |
| CYLD (AAH12342, 854 a.a. ∼ 953 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence | |
| 3A9 | |
| Mouse monoclonal antibody raised against a partial recombinant CYLD. | |
| CYLD | |
| CDMT/CYLD1/CYLDI/EAC/FLJ20180/FLJ31664/FLJ78684/HSPC057/KIAA0849/MFT/MFT1/SBS/TEM/USPL2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 1540 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.