Learn More
Cyclin D1 Mouse anti-Human, FITC, Clone: G124-326, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554109
Includes: FITC Mouse Anti-Human Cyclin D1 and FITC Mouse IgG1, κ Isotype Control
Description
Cyclins and cyclin-dependent kinases (cdks) are subunits of cell cycle dependent protein kinases that regulate key events during the progression of the cell cycle. Specific substrates for cdk/cyclin kinases include nuclear lamins, histones, oncogenes (c-src, c-abl, SV40 large-T), tumor suppressor genes (the retinoblastoma protein [Rb] and p53), nucleolin, RNA polymerase II and others. Human cyclin D1 migrates at a molecular weight of 36kDa in SDS/PAGE.
Clone G124-326 recognizes human Cyclin D1. It does not cross-react with human cyclins D2 and D3. Recombinant full-length human Cyclin D1 was used as immunogen. The G124-326 antibody was initially characterized by immunoprecipitation and western blot analysis. In publications it has been used for western blot analysis, flow cytometric analysis, and immunohistochemistry of frozen tissue sections.
Clone MOPC-21 is a mouse IgG1 isotype (negative) control. The MOPC-21 antibody has unknown specificity. The G124-326 and MOPC-21 FITC conjugates are matched in F/P ratios. The optimum F/P ratio was experimentally determined by flow cytometric analysis.
Host Species: Mouse
Clone: G124-326, MOPC-21
Isotype: IgG1, IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| Cyclin D1 | |
| Monoclonal | |
| FITC | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| G124-326 | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.