Learn More
Cyclin A Mouse anti-Human, Unlabeled, Clone: BF683, BD
Mouse Monoclonal Antibody
£193.97 - £312.42
Specifications
| Antigen | Cyclin A |
|---|---|
| Clone | BF683 |
| Concentration | 0.5mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15867598
|
BD Biosciences
554174 |
0.1 mg |
£202.00 £193.97 / Each Save £8.03 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15877598
|
BD Biosciences
554175 |
0.25 mg | |||||||
Description
Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins (regulatory subunits) bind to cdks (catalytic subunits) to form complexes that regulate the progression of the cell cycle. The main cyclin-cdks complexes formed in vertebrate cells are cyclin D-cdk4 (G0/G1), cyclin E-cdk2 (G1/S), cyclin A-cdk2 (S) and cyclin B1-cdk1 (G2/M). These complexes are regulated by activating and inhibitory phosphorylation events, as well as by interactions with small regulatory proteins including p21 and p27Kip1. Specific substrates for cdk-cyclin complexes include nuclear lamins, histones, oncogenes (e.g., c-abl and SV40 large T-Ag), tumor suppressor genes (e.g., retinoblastoma protein, Rb), nucleolin and others. Cyclin A is involved in both S-phase and G2/M transitions of the cell cycle through its association with cdk2 and cdk1, respectively. Cyclin A may also form a complex with the adenovirus oncoprotein E1A which has DNA binding activity. Human cyclin A has been reported to migrate between 54-60kDa by SDS-PAGE and clone BF683 reportedly does not cross-react with mouse, rat or mink cyclin A.
Host Species: Mouse
Clone: BF683
Isotype: IgE
Species Reactivity: Human
Immunogen: Human Cyclin A Recombinant Protein
Formula Weight [Chemical]: 54-60kDa
Flow Cytometry, Immunoprecipitation, Western Blotting
Specifications
| Cyclin A | |
| 0.5mg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgE | |
| Affinity Purified |
| BF683 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human Cyclin A Recombinant Protein | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.