missing translation for 'onlineSavingsMsg'
Learn More

Cyclin A Mouse anti-Human, Unlabeled, Clone: 25, BD

Product Code. 12820510
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12820510 50 μg Each
15835859 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12820510 Supplier BD Biosciences Supplier No. 611268

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Progression of the mammalian cell cycle is regulated by phosphorylation of many key proteins. Several classes of cyclins (A-E) act as regulatory subunits for cyclin-dependent kinases (cdks). These cyclin-cdk holoenzymes are essential for proper control of cell cycle progression. They phosphorylate and regulate a variety of substrates whose activity is required for cell cycle transitions. The temporal expression of cyclins is tightly regulated throughout the cell cycle by synthesis and degradation. Such regulation plays a critical role in controlling the enzymatic activity of the cdks. Cyclin A, one of the mitotic cyclins, activates Cdk2 near the start of S phase and is necessary for the initiation of DNA replication. In mammalian somatic cells, Cyclin A is required during S phase and passage through G2. The D and E type cyclins regulate passage through G1, while Cyclin B is a critical regulator of mitosis. It has been shown in a number of species that mutation or disruption of normal Cyclin A expression causes cells to arrest at G2. Cyclin A binds both the cdc2 (Cdk1) and Cdk2 kinases and may also have a role in mitotic dependence on S phase completion.

Host Species: Mouse
Clone: 25
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Cyclin A aa. 26-144
Formula Weight [Chemical]: 60kDa

Bioimaging, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A
Applications Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Cyclin A aa. 26-144
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.