missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cytochrome b5 reductase 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CYB5R2 protein.
Brand: Abnova H00051700-B02P.50ug
This item is not returnable.
View return policy
Description
Sequence: MNSRRREPITLQDPEAKYPLPLIEKEKISHNTRRFRFGLPSPDHVLGLPVGNYVQLLAKIDNELVVRAYTPVSSDDDRGFVDLIIKIYFKNVHPQYPEGGKMTQYLENMKIGETIFFRGPRGRLFYHGPGNLGIRPDQTSEPKKTLADHLGMIAGGTGITPMLQLIRHITKDPSDRTRMSLIFANQTEEDILVRKELEEIARTHPDQFDLWYTLDRPPIGPWSAEGATLLSNSAQFHSpecifications
| cytochrome b5 reductase 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human CYB5R2 protein. | |
| CYB5R2 | |
| B5R.2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 51700 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC001346.1 | |
| CYB5R2 | |
| CYB5R2 (AAH01346.1, 1 a.a. ∼ 237 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction