missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CXCR6, PerCP, Clone: 221002, R&D Systems™
Rat Monoclonal Antibody
Specifications
| Antigen | CXCR6 |
|---|---|
| Clone | 221002 |
| Immunogen | CHO Chinese hamster ovary cell line transfected with mouse CXCR6 Met1-Leu351 Accession # Q9EQ16 |
| Host Species | Rat |
| Primary or Secondary | Primary |
Description
CXCR6 Monoclonal specifically detects CXCR6 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CXCR6 | |
| CHO Chinese hamster ovary cell line transfected with mouse CXCR6 Met1-Leu351 Accession # Q9EQ16 | |
| Primary | |
| Mouse | |
| PerCP | |
| 10663 | |
| Purified | |
| Flow Cytometry |
| 221002 | |
| Rat | |
| Monoclonal | |
| IgG2b | |
| Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Protein A or G purified from hybridoma culture supernatant | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title