Learn More
chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1), Mouse, Clone: 1F10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CXCL12.
Brand: Abnova H00006387-M03.100ug
Description
For background information on chemokines, see CXCL1 (MIM 155730). Stromal cell-derived factors 1-alpha and 1-beta are small cytokines that belong to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF (see MIM 191160), or IL1 (see MIM 147760). The intercrines are characterized by the presence of 4 conserved cysteines which form 2 disulfide bonds. They can be classified into 2 subfamilies. In the CC subfamily, which includes beta chemokine, the cysteine residues are adjacent to each other. In the CXC subfamily, which includes alpha chemokine, they are separated by an intervening amino acid. The SDF1 proteins belong to the latter group.[supplied by OMIM
Sequence: MNAKVVVVLVLVLTALCLSDGKPVSLSYRCPCRFFESHVARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKALNKSpecifications
| chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC039893 | |
| CXCL12 | |
| CXCL12 (AAH39893.1, 1 a.a. ∼ 89 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1F10 | |
| Mouse monoclonal antibody raised against a full length recombinant CXCL12. | |
| CXCL12 | |
| PBSF/SCYB12/SDF-1a/SDF-1b/SDF1/SDF1A/SDF1B/TLSF-a/TLSF-b/TPAR1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6387 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.