missing translation for 'onlineSavingsMsg'
Learn More

anti-CTLA-4/CD152, Polyclonal, Invitrogen™

CTLA-4/CD152 Polyclonal antibody PA5-26465 from Thermo Scientific™ specifically detects CTLA-4/CD152 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence and Western Blot.

Brand:  Invitrogen PA5-26465

Product Code. 13221479

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CTLA-4/CD152 Polyclonal antibody PA5-26465 from Thermo Scientific™ specifically detects CTLA-4/CD152 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence and Western Blot.

CTLA4 is a member of the immunoglobulin superfamily and encodes a protein which transmits an inhibitory signal to T cells. The protein contains a V domain, a transmembrane domain, and a cytoplasmic tail. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The membrane-bound isoform functions as a homodimer interconnected by a disulfide bond, while the soluble isoform functions as a monomer.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: KLH conjugated synthetic peptide between 182-210 amino acids from the C-terminal region of human CTLA4
Formulation: 400μL Purified Antibody (Lot-specific) in PBS, with 0.09% sodium azide
Synonyms: CELIAC3, CTLA-4, GRD4, GSE, ICOS, IDDM12, CD152 isoform, celiac disease 3, cytotoxic T lymphocyte associated antigen 4 short spliced form, cytotoxic T-lymphocyte antigen 4
Uniprot Accession: P16410
Entrez GeneID: 1493
Gene Symbol: CTLA4
This antibody is predicted to react with mouse, porcine and rabbit based on sequence homology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CTLA-4/CD152
Polyclonal
P16410
CTLA4
KLH conjugated synthetic peptide between 182-210 amino acids from the C-terminal region of human CTLA4
RUO
1493
Store at 2°-8°C for up to 6 months. For long term storage store at -20°C
IgG
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CELIAC3, CTLA-4, GRD4, GSE, ICOS, IDDM12, CD152 isoform, celiac disease 3, cytotoxic T lymphocyte associated antigen 4 short spliced form, cytotoxic T-lymphocyte antigen 4
Rabbit
400 μL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.