Learn More
CRMP1, Mouse, Clone: 2B6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CRMP1.
Brand: Abnova H00001400-M50.100ug
Description
This gene encodes a member of a family of cytosolic phosphoproteins expressed exclusively in the nervous system. The encoded protein is thought to be a part of the semaphorin signal transduction pathway implicated in semaphorin-induced growth cone collapse during neural development. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: HVVPEPGSSLLTSFEKWHEAADTKSCCDYSLHVDITSWYDGVREELEVLVQDKGVNSFQVYMAYKDVYQMSDSQLYEAFTFLKGLGAVILSpecifications
| CRMP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001014809 | |
| CRMP1 | |
| CRMP1 (NP_001014809.1, 221 a.a. ∼ 310 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2B6 | |
| Mouse monoclonal antibody raised against a partial recombinant CRMP1. | |
| CRMP1 | |
| DPYSL1/DRP-1/DRP1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1400 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.