Learn More
CRHBP, Human, IP-WB Antibody Pair (PW3), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00001393-PW3.1Set
Product Code. 16171914
  • £729.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-CRHBP (300μL), Antibody pair for WB: mouse purified polyclonal anti-CRHBP (50μg)
Specifications
CRHBP
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
CRF-BP/CRFBP
Mouse, Rabbit
RUO
1393
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
CRHBP
CRHBP
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-CRHBP (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-CRHBP (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.