missing translation for 'onlineSavingsMsg'
Learn More

cAMP responsive element binding protein 5, Mouse, Clone: 8A5, Abnova™

Mouse monoclonal antibody raised against a partial recombinant CREB5.

Brand:  Abnova H00009586-M02.100ug

Product Code. 16180986

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: MFCTSGGNSASVMSMRPVPGSLSSLLHLHNRQRQPMPASMPGTLPNPTMPGSSAVLMPMERQMSVNSSIMGMQGPNLSNPCASPQVQPMHSEAKMRLKA
Specifications

Specifications

cAMP responsive element binding protein 5
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_001011666
CREB5
CREB5 (NP_001011666, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Transcription Regulation
Primary
Human, Mouse
Antibody
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
8A5
Mouse monoclonal antibody raised against a partial recombinant CREB5.
CREB5
CRE-BPA
Mouse
Affinity Purified
RUO
Yes
9586
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.