missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREB3L2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA537966
This item is not returnable.
View return policy
Description
This antibody is predicted to react with human, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).
Specifications
| CREB3L2 | |
| Polyclonal | |
| Unconjugated | |
| Creb3l2 | |
| basic transcription factor 2; BBF2 human homolog on chromosome 7; BBF2H7; B-ZIB transcription factor; C530025K05Rik; cAMP responsive element binding protein 3 like 2; cAMP responsive element binding protein 3-like 2; cAMP-responsive element-binding protein 3-like protein 2; Creb3l2; Cyclic AMP-responsive element-binding protein 3-like protein 2; FUS/BBF2H7 protein; Processed cyclic AMP-responsive element-binding protein 3-like protein 2; Ra69; SCI-related protein Ra69; SCIRR69; TCAG_1951439 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 208647, 362339, 64764 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6QDP7, Q70SY1, Q8BH52 | |
| Creb3l2 | |
| Peptide with sequence C-HSLQEPYTASVVRS, from the internal region of CREB3L2 (aa 413-426). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction