Learn More
CRACC (CD319) Mouse anti-Human, Alexa Fluor 647, Clone: 235614, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564338
Description
The 235614 monoclonal antibody specifically binds to CRACC (CD2-like receptor activating cytotoxic cells) which is also known as CD319, CS1 (CD2 subset 1), SLAMF7, 19A, and 19A24. CRACC is a type I transmembrane glycoprotein that belongs to the CD2 family of receptors within the Ig superfamily. CRACC is expressed on most NK cells and subsets of CD8+ T cells, CD4+ T cells, B cells, macrophages, and dendritic cells. CRACC reportedly plays roles in the activation, proliferation and effector functions of T cells, NK cells, and other leucocytes.
Specifications
| CRACC (CD319) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| 235614 | |
| CRACC; SLAMF7; 19A; 19A24 protein; CS1 | |
| Human CRACC/SLAMF7 | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.