Learn More
anti-COX IV, Polyclonal, Abcam™
Brand: Abcam AB16056-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human, Xenopus laevis, Potato, African Green Monkey, Chinese Hamster Predicted to work with Chimpanzee
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 150 to the C-terminus of Human COX IV.
Applications:
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
Specifications
| COX IV | |
| 0.8-1mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| IgG |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 150 to the C-terminus of Human COX IV. | |
| 100 μg | |
| Chimpanzee, Hamster, Human, Monkey, Mouse, Potato, Rat, Xenopus species |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.