missing translation for 'onlineSavingsMsg'
Learn More

Cortactin Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

£441.00

Specifications

Antigen Cortactin
Clone 30
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15835229
View Documents
BD Biosciences
610050
150 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Cortactin is a protein which is represented by two structurally related isoforms of 80 kDa and 85 kDa. Sequence analysis of this protein reveals the presence of a carboxy-terminal SH3 domain, as well as six internal tandem repeats of a 37 amino acid sequence. It is able to bind to F-actin via this N-terminal repeat region, and colocalizes with actin in the cell. Cortactin was originally described as a protein that is tyrosine-phosphorylated in cells transformed by the src oncogene, in which it localizes to rosettes or podosomes. It has since been shown that other stimuli, such as bacterial invasion, treatment with growth factors, and integrin-mediated cell adhesion, are also capable of inducing phosphorylation of cortactin. Some additional evidence indicates that cortactin may actually be a substrate of syk, with src acting as an upstream signaling molecule.
Specifications

Specifications

Cortactin
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
30
Western Blot
Unconjugated
RUO
Canine, Human
Chicken Cortactin
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.