Learn More
coronin, actin binding protein, 1C, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CORO1C.
Brand: Abnova H00023603-A01.50uL
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. [provided by RefSeq
Sequence: EEWFEGKNADPILISLKHGYIPGKNRDLKVVKKNILDSKPTANKKCDLISIPKKTTDTASVQNEAKLDEILKEIKSIKDTICNQDERISKLEQQMAKIASpecifications
| coronin, actin binding protein, 1C | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CORO1C. | |
| CORO1C | |
| HCRNN4 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 23603 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_014325 | |
| CORO1C | |
| CORO1C (NP_055140, 375 a.a. ∼ 473 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.