missing translation for 'onlineSavingsMsg'
Learn More

coatomer protein complex, subunit epsilon, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant COPE.

Brand:  Abnova H00011316-A01.50uL

Product Code. 16174066

  • £209.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi nonclathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: KCSPTLLLLNGQAACHMAQGRWEAAEGLLQEALDKDSGYPETLVNLIVLSQHLGKPPEVTNRYLSQLKDAHRSHPFIKEYQAKENDFDRLVLQYAPSA
Specifications

Specifications

coatomer protein complex, subunit epsilon
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant COPE.
COPE
FLJ13241/epsilon-COP
Mouse
50 μL
Yes
11316
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
NM_007263
COPE
COPE (NP_009194, 211 a.a. ∼ 308 a.a) partial recombinant protein with GST tag.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.