Learn More
Invitrogen™ Complement C5a Monoclonal Antibody (2952)
GREENER_CHOICE
Mouse Monoclonal Antibody
Marque:  Invitrogen™ MA140162
Code produit. 11539702
  • £572.00 / 100µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
MA1-40162 detects C5a/C5a des-Arg from human samples. MA1-40162 has been successfully used in ELISA, Immunohistochemsitry (Frozen, Paraffin), Functional Assays and Western blot applications. The MA1-40162 immunogen is the Neo-epitope on C5a. The MA1-40162 immunogen is the Neo-epitope on C5a (ISHKDMQLG amino acid 65-73). Endotoxin <24 EU/mg. Positive control Human Granulocytes. General comment: Please notice that under given conditions it is known that C5 can expose epitopes normally only found in the cleaved activation products.
The protein encoded by this gene is the fifth component of complement, which plays an important role in inflammatory and cell killing processes. This protein is comprised of alpha and beta polypeptide chains that are linked by a disulfide bridge. An activation peptide, C5a, which is an anaphylatoxin that possesses potent spasmogenic and chemotactic activity, is derived from the alpha polypeptide via cleavage with a convertase. The C5b macromolecular cleavage product can form a complex with the C6 complement component, and this complex is the basis for formation of the membrane attack complex, which includes additional complement components. Mutations in this gene cause complement component 5 deficiency, a disease where patients show a propensity for severe recurrent infections. Defects in this gene have also been linked to a susceptibility to liver fibrosis and to rheumatoid arthritis.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Complement C5a
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and no preservative
P01031
C5
Neo-epitope on C5a.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
2952
Unconjugated
C5
anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905
Mouse
Protein G
RUO
727
4°C
Liquid
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis