missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen II Monoclonal Antibody (2B1.5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512789
Code : LE
Additional Details : Weight : 0.01000kg
Description
Staining of formalin/parafin tissues requires disgestion of tissue sections with pepsin at 1 mg/mL Tris-HCl, pH 2.0 for 15 min at RT or 10 min at 37C.
Collagen II Monoclonal antibody specifically detects Collagen II in Bovine, Chicken, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Collagen II | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02458, P02459, P02460, P05539, P28481 | |
Col2a1 | |
A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
500 μL | |
Primary | |
Bovine, Chicken, Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2B1.5 | |
Unconjugated | |
Col2a1 | |
alpha 1 type II procollagen; alpha 1 type IIA collagen; alpha 1 type IIA collagen precursor; alpha collagen type II; alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CB11; CG2A1A; CGA; Chondrocalcin; COL11A3; Col2; Col2a; col2a1; Col2a-1; col2a1.L; col2a1a; col2a1-a; col2a1-b; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen type II alpha 1 chain; collagen type IIA; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); collagen, type II, alpha 1 L homeolog; COLLII; cyanogen bromide; Del1; disproportionate micromelia; Dmm; Lpk; M100856; MGC131516; pro alpha1 type II collagen; pro-alpha 1(II); pro-alpha-1 type II collagen; procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; Rgsc856; SEDC; STL1; type II collagen; type II collagen cyanogen bromide fra | |
Mouse | |
Protein A | |
RUO | |
1280, 12824, 25412, 395069, 407142 | |
4° C | |
Liquid |