missing translation for 'onlineSavingsMsg'
Learn More

Cofilin Mouse, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

£237.00 - £284.36

Specifications

Antigen Cofilin
Clone 32
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15856109
View Documents
BD Biosciences
612144
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15866109
View Documents
BD Biosciences
612145
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cell motility is a basic cellular behavior involved in embryogenesis, neurite outgrowth, wound healing, inflammation, and cancer metastasis. Filamentous actin is an essential component of cell protrusions involved in cell motility. The protrusion formation is regulated by actin polymerization and depolymerization. Cofilin is a ubiquitously expressed G- and F-actin binding protein that contains a nuclear localization signal, a C-terminal hexapeptide sequence that is identical to tropomyosin, and other regions that are homologous to actin binding proteins. In vitro, cofilin has actin severing activity and induces an increased off-rate from the pointed end of actin filaments. These activities can increase the rate of actin polymerization and depolymerization, as well as increase the number of barbed ends available for polymerization. Stimulation of MTLn3 cells with EGF leads to an increase in cofilin at the leading edge of laemellipodia, which correlates with an increase in the number of barbed ends at the leading edge. Cofilin function blocking antibodies inhibit both the appearance of these barbed ends and lamellipodial protrusion. Thus, cofilin is an actin binding protein that regulates actin filament formation during cell motility.

Western Blotting

Specifications

Specifications

Cofilin
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
32
Western Blot
Unconjugated
RUO
Canine, Human, Mouse, Rat
Mouse Cofilin aa. 3-98
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.