Learn More
contactin associated protein-like 4, Mouse, Clone: 5G1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CNTNAP4.
Marca: Abnova H00085445-M01A.200uL
Descrizione
This gene product belongs to the neurexin family, members of which function in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, and thrombospondin N-terminal-like domains. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: VLGRILEHSDVDQETALAGAQGFTGCLSAVQLSHVAPLKAALHPSHPDPVTVTGHVTESSCMAQPGTDATSRERTHSFADHSGTIDDREPLANAIKSDSASpecifica
| contactin associated protein-like 4 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_033401 | |
| CNTNAP4 | |
| CNTNAP4 (NP_207837, 1145 a.a. ∼ 1244 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Yes | |
| 85445 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5G1 | |
| Mouse monoclonal antibody raised against a partial recombinant CNTNAP4. | |
| CNTNAP4 | |
| CASPR4/KIAA1763 | |
| Mouse | |
| 200 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.