Learn More
calponin 3, acidic, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CNN3.
Brand: Abnova H00001266-A01.50uL
Description
This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction. [provided by RefSeq
Sequence: DQKLTLQPVDNSTISLQMGTNKVASQKGMSVYGLGRQVYDPKYCAAPTEPVIHNGSQGTGTNGSEISDSDYQAEYPDEYHGEYQDDYPRDYQYSDQGIDYSpecifications
| calponin 3, acidic | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CNN3. | |
| CNN3 | |
| CNN3 | |
| CNN3 (NP_001830, 230 a.a. ∼ 329 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001839 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 1266 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.