Learn More
Clusterin Mouse anti-Human, Unlabeled, Clone: E5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 552886
Description
The E5 antibody reacts with human clusterin. Clusterin was originally isolated and characterized from glomerular immune deposits of patients with renal glomerulonephritis. Clusterin, an 80kDa disulfide-linked, heterodimeric glycoprotein (previously named SP-40,40 [serum protein 40kDa,40kDa]) was shown to be part of the SC5b-9 complex of complement, yet separate and distinct from the S protein. Clusterins have since been identified in a number of mammalian species and have been implicated in diverse cellular activities, however, the exact role remains to be elucidated.
Host Species: Mouse
Clone: E5
Isotype: IgG1 κ
Species Reactivity: Human
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| Clusterin | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 1 mL | |
| Cell Biology | |
| Human | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| E5 | |
| Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.